Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS4 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299438FR
Description
ADAMTS4 Polyclonal specifically detects ADAMTS4 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ADAMTS4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 4, ADAM metallopeptidase with thrombospondin type 1 motif, 4, ADAM-TS 4, ADAMTS-2, ADAM-TS4, ADAMTS-4, ADMP-1EC 3.4.24.82, aggrecanase-1, EC 3.4.24, KIAA0688A disintegrin and metalloproteinase with thrombospondin motifs 4 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse ADAMTS4. | |
0.1 mL | |
Cancer, GPCR, Neuroscience, Vision | |
9507 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction