Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ADAMTS4 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ADAMTS4 Polyclonal specifically detects ADAMTS4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ADAMTS4 | |
Polyclonal | |
Rabbit | |
Cancer, GPCR, Neuroscience, Vision | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 4, ADAM metallopeptidase with thrombospondin type 1 motif, 4, ADAM-TS 4, ADAMTS-2, ADAM-TS4, ADAMTS-4, ADMP-1EC 3.4.24.82, aggrecanase-1, EC 3.4.24, KIAA0688A disintegrin and metalloproteinase with thrombospondin motifs 4 | |
ADAMTS4 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
9507 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VEGLTVQYLGQAPELLGGAEPGTYLTGTINGDPESVASLHWDGGALLGVLQYRGAELHLQPLEGGTPNSAGGPGAHILRRKSPASGQGPMCN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title