Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTS4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585211
Description
Keep as concentrated solution. Predicted reactivity: Mouse (94%), Rat (94%), Bovine (94%). Positive Control: mouse brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the ADAMTS protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene lacks a C-terminal TS motif. It is responsible for the degradation of aggrecan, a major proteoglycan of cartilage, and brevican, a brain-specific extracellular matrix protein. The cleavage of aggrecan and brevican suggests key roles of this enzyme in arthritic disease and in the central nervous system, potentially, in the progression of glioma.
Specifications
ADAMTS4 | |
Polyclonal | |
Unconjugated | |
ADAMTS4 | |
3830423K05; a disintegrin and metallopeptidase with thrombospondin motifs 4; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 4; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 4; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 4; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 4; ADAM metallopeptidase with thrombospondin type 1 motif, 4; ADAMs; ADAM-TS 4; ADAMTS family member; ADAMTS-2; Adamts4; ADAM-TS4; ADAMTS-4; ADMP-1; Aggrecanase; Aggrecanase 1; aggrecanase-1; disintegrin and metalloproteinase with thrombospondin motifs 2; KIAA0688; metalloendopeptidases; metalloproteinase; mKIAA0688; UNQ769/PRO1563 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
240913, 9507 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O75173, Q8BNJ2 | |
ADAMTS4 | |
Recombinant protein encompassing a sequence within the center region of human ADAMTS4. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction