Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | ADAMTS5 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ADAMTS5 Polyclonal specifically detects ADAMTS5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ADAMTS5 | |
Polyclonal | |
Rabbit | |
GPCR | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
11096 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KSTPKVNSVTSHGSNKVGSHTSQPQWVTGPWLACSRTCDTGWHTRTVQCQDGNRKLAKGCPLSQRPSAFKQCLLKKC | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
A disintegrin and metalloproteinase with thrombospondin motifs 11, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 5 (aggrecanase-2), ADAM metallopeptidase with thrombospondin type 1 motif, 5, ADAM-TS 11, ADAM-TS 5, ADAMTS-11, ADAM-TS5, ADAMTS-5, ADMP2, ADMP-2A disintegrin and metalloproteinase with thrombospondin motifs 5, Aggrecanase-2, disintegrin-like and metalloprotease with thrombospondin type 1 motif, 510ADAMTS11FLJ36738, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82 | |
ADAMTS5 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title