Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA514350
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA514350 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA514350 Supplier Invitrogen™ Supplier No. PA514350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAMTS5 Polyclonal Antibody for Western Blot

ADAMTS5, also known as Implantin or Aggrecanase 2, is a member of the larger family of ADAMs (A Disintegrin And Metalloproteinase) metalloproteinases containing thrombospondin (TS) repeats. ADAMTS5 (A Disintegrin And Metalloproteinase with Thrombospondin-5 motif) was first described as ADAMTS5, a protein elevated in mice during the peri-implantion period. At the same time, another group identified Aggrecanase 11, a protein elevated in arthritic synovium. The name was later changed to ADAMTS5. ADAMTS5 is expressed in human and mouse. It has been found in heart, lung, cervix, uterus, ovary, brain, cartilage, and numerous other tissues, as well as chondroblastoma cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADAMTS5
Gene Accession No. Q9R001, Q9UNA0
Gene Alias 9530092O11Rik; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 11; A disintegrin and metalloproteinase with thrombospondin motifs 5; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 5; ADAM metallopeptidase with thrombospondin type 1 motif, 5; ADAMs; ADAM-TS 11; ADAM-TS 5; ADAMTS family member; ADAMTS1; ADAMTS11; ADAMTS-11; ADAMTS5; ADAM-TS5; ADAMTS-5; ADMP2; ADMP-2; Aggrecanase 2; Aggrecanase-2; AI481094; ASMP-2; Implantin; metalloendopeptidases
Gene Symbols ADAMTS5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 331-360 amino acids from the central region of human ADAMTS5.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11096, 23794
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.