Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTS6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560365
Description
Immunogen sequence: LHGVIATEDE EYFIEPLKNT TEDSKHFSYE NGHPHVIYKK SALQQRHLYD HSHCGVSDFT RSGKPWWLND TSTVSYSLPI NNTHIHHRQK RSVSIERFVE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The encoded preproprotein is proteolytically processed to generate the mature enzyme. Expression of this gene may be regulated by the cytokine TNF-alpha. [provided by RefSeq, Mar 2016].
Specifications
ADAMTS6 | |
Polyclonal | |
Unconjugated | |
ADAMTS6 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 6; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 6; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 6; ADAM metallopeptidase with thrombospondin type 1 motif, 6; ADAMs; ADAM-TS 6; ADAMTS6; ADAM-TS6; ADAMTS-6; metalloendopeptidases | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11174 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKP5 | |
ADAMTS6 | |
Recombinant protein corresponding to Human ADAMTS6. Recombinant protein control fragment (Product #RP-102943). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction