Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114171 Supplier Invitrogen™ Supplier No. PA5114171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PWSECSATCA GGVQRQEVVC KRLDDNSIVQ NNYCDPDSKP PENQRACNTE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The encoded preproprotein is proteolytically processed to generate the mature enzyme. Expression of this gene may be regulated by the cytokine TNF-alpha. [provided by RefSeq, Mar 2016].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADAMTS6
Gene Accession No. Q9UKP5
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 6; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 6; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 6; ADAM metallopeptidase with thrombospondin type 1 motif, 6; ADAMs; ADAM-TS 6; ADAMTS6; ADAM-TS6; ADAMTS-6; metalloendopeptidases
Gene Symbols ADAMTS6
Host Species Rabbit
Immunogen Immunogen sequence: PWSECSATCA GGVQRQEVVC KRLDDNSIVQ NNYCDPDSKP PENQRACNTE.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11174
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.