Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565516 Supplier Invitrogen™ Supplier No. PA565516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WDLQTVAVWGT FLPTTLTGLG HTPEPALNPG PKGQPESLSP EVPLSSRLLS M Highest antigen sequence identity to the following orthologs - mouse 35%, rat 36%.

ADAMTS (a disintegrin and metalloproteinase domain with thrombospondin type-1 modules) is a family of zinc-dependent proteases that are implicated in a variety of normal and pathological conditions, including arthritis and cancer. ADAMTS protein family members contain an N-terminal propeptide domain, a metalloproteinase domain, a disintegrin-like domain and a C-terminus that contains a varying number of thrombospondin type-1 (TSP-1) motifs. ADAMTS genes are primarily expressed in fetal tissues, including lung, kidney and liver. ADAMTS-7 (ADAM metallopeptidase with thrombospondin type 1 motif, 7), also known as COMPase, is a 1,686 amino acid protein that exists as two alternatively spliced isoforms. Encoded by a gene that maps to human chromosome 15q25.1, ADAMTS-7 contains eight TSP-1 motifs and binds one zinc ion per subunit. ADAMTS-7 is expressed in heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas. ADAMTS-7 is also located in meniscus, bone, tendon, cartilage, synovium, fat and ligaments, and is up-regulated in articular cartilage and synovium in arthritis patients. ADAMTS-7 functions as a metalloprotease and may play a role in the degradation of COMP. ADAMTS-7 is pH dependent, with optimum pH between 7.5 and 9.5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAMTS7
Gene Accession No. Q9UKP4
Gene Alias a disintegrin and metalloprotease with thrombospondin motifs-7 preproprotein; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 7; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 7; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 7; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 7; ADAM metallopeptidase with thrombospondin type 1 motif, 7; ADAMs; ADAM-TS 7; ADAMTS7; ADAM-TS7; ADAMTS-7; ADAMTS7B; COMPase; metalloendopeptidases; metalloprotease
Gene Symbols ADAMTS7
Host Species Rabbit
Immunogen Recombinant Human ADAMTS7. Recombinant protein control fragment (Product #RP-106192).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11173
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.