Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS7 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980271
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0271 Supplier Bioss Supplier No. BS0433R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAMTS (a disintegrin and metalloproteinase domain with thrombospondin type-1 modules) is a family of zinc-dependent proteases that are implicated in a variety of normal and pathological conditions, including arthritis and cancer. ADAMTS protein family members contain an N-terminal propeptide domain, a metalloproteinase domain, a disintegrin-like domain and a C-terminus that contains a varying number of thrombospondin type-1 (TSP-1) motifs. ADAMTS genes are primarily expressed in fetal tissues, including lung, kidney and liver. ADAMTS-7 (ADAM metallopeptidase with thrombospondin type 1 motif, 7), also known as COMPase, is a 1,686 amino acid protein that exists as two alternatively spliced isoforms. Encoded by a gene that maps to human chromosome 15q25.1, ADAMTS-7 contains eight TSP-1 motifs and binds one zinc ion per subunit. ADAMTS-7 is expressed in heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas. ADAMTS-7 is also located in meniscus, bone, tendon, cartilage, synovium, fat and ligaments, and is up-regulated in articular cartilage and synovium in arthritis patients. ADAMTS-7 functions as a metalloprotease and may play a role in the degradation of COMP. ADAMTS-7 is pH dependent, with optimum pH between 7.5 and 9.5.

Specifications

Antigen ADAMTS7
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADAMTS7
Gene Accession No. Q9UKP4
Gene Alias a disintegrin and metalloprotease with thrombospondin motifs-7 preproprotein; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 7; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 7; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 7; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 7; ADAM metallopeptidase with thrombospondin type 1 motif, 7; ADAMs; ADAM-TS 7; Adamts7; ADAM-TS7; ADAMTS-7; ADAMTS7B; COMPase; metalloendopeptidases; metalloprotease
Gene Symbols ADAMTS7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ADAMTS7.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11173
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.