Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS8 Antibody (OTI2H5), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271677JF646
Description
ADAMTS8 Monoclonal specifically detects ADAMTS8 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ADAMTS8 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADAMTS8 | |
Full length recombinant protein of human ADAMTS8 (NP_008968) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2H5 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 8, ADAM metallopeptidase with thrombospondin type 1 motif, 8, ADAMTS-8, ADAM-TS8A disintegrin and metalloproteinase with thrombospondin motifs 8, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82, FLJ41712, METH-2, METH2ADAM-TS 8, METH-8 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
11095.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction