Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS8 Monoclonal Antibody (OTI2H5)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526187

Catalog No. PIMA526187


Only null left
Add to Cart

Description

Description

ADAMTS8 Monoclonal Antibody for Western Blot, ICC/IF

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains two C-terminal TS motifs, and disrupts angiogenesis in vivo. A number of disorders have been mapped in the vicinity of this gene, most notably lung neoplasms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ADAMTS8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UP79
ADAMTS8
Full length human recombinant protein of ADAMTS8 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunocytochemistry
OTI2H5
Unconjugated
ADAMTS8
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 8; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 8; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 8; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 8; ADAM metallopeptidase with thrombospondin type 1 motif, 8; ADAMs; ADAM-TS 8; ADAMTS8; ADAM-TS8; ADAMTS-8; ADAMTS-8 precursor (A disintegrin and metalloproteinase with thrombospondin motifs 8) (ADAM-TS 8) (ADAM-TS8) (METH-2); EC 3.4.24.14; EC 3.4.24.82; FLJ41712; metalloendopeptidases; METH2; METH-2; METH2ADAM-TS METH-8; METH-8
Mouse
Affinity chromatography
RUO
11095
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.