Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986863
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6863 Supplier Bioss Supplier No. BS5859R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains two C-terminal TS motifs, and disrupts angiogenesis in vivo. A number of disorders have been mapped in the vicinity of this gene, most notably lung neoplasms.

Specifications

Antigen ADAMTS8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADAMTS8
Gene Accession No. Q9UP79
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 8; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 8; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 8; ADAM metallopeptidase with thrombospondin type 1 motif, 8; ADAMs; ADAM-TS 8; ADAMTS8; ADAM-TS8; ADAMTS-8; EC 3.4.24.14; EC 3.4.24.82; FLJ41712; metalloendopeptidases; METH2; METH-2; METH2ADAM-TS METH-8; METH-8
Gene Symbols ADAMTS8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ADAMTS8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11095
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.