Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074345
Description
ADAMTS9 Polyclonal specifically detects ADAMTS9 in Human samples. It is validated for Western Blot.Specifications
ADAMTS9 | |
Polyclonal | |
Unconjugated | |
PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
ADAM metallopeptidase with thrombospondin type 1 motif, 9, ADAMTS-9,9, FLJ42955 | |
Rabbit | |
Affinity Purified | |
RUO | |
56999 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
1 mg/mL | |
Western Blot 0.5 ug/mL | |
Q9P2N4 | |
ADAMTS9 | |
Synthetic peptide corresponding to residues C F D G K H F N I N G L L P N of human ADAMTS9. | |
100 μg | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction