Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTSL1 Monoclonal Antibody (1B2-8-2)

Catalog No. PIMA533368
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533368 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533368 Supplier Invitrogen™ Supplier No. MA533368

Mouse Monoclonal Antibody

Product is supplied at 1 mL (100 μL/mL). Western blotting applications the antibody works better in non-reducing than reducing conditions although it will also recognize the reduced antigen at correspondingly higher concentrations.

Punctin (ADAMTSL-1) is a secreted molecule, expressed in adult skeletal muscle, resembling members of the ADAMTS family of proteases, containing four thrombospondin type I repeats. Human punctin cleaves aggrecan and is thereby classified as an aggrecanase. Like the ADAMTS proteases, each domain in punctin has an even number of cysteine residues suggesting that each domain may have internal disulfide bonds and that punctin consists of a series of independently folded and disulfide-bonded domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTSL1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B2-8-2
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene ADAMTSL1
Gene Accession No. Q8N6G6
Gene Alias 5930437A14Rik; 6720426B09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 1; ADAM-TS related protein 1; Adamtsl1; ADAMTSL-1; ADAMTS-like 1; ADAMTS-like protein 1; ADAMTSR1; C9orf94; DKFZp686L03130; FLJ35283; FLJ41032; FLJ46891; metalloendopeptidases; MGC118803; MGC118805; MGC40193; PUNCTIN; Punctin-1; RP11-220B22.2; UNQ528/PRO1071
Gene Symbols ADAMTSL1
Host Species Mouse
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92949
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.