Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTSL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531013
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Punctin (ADAMTSL-1) is a secreted molecule, expressed in adult skeletal muscle, resembling members of the ADAMTS family of proteases, containing four thrombospondin type I repeats. Human punctin cleaves aggrecan and is thereby classified as an aggrecanase. Like the ADAMTS proteases, each domain in punctin has an even number of cysteine residues suggesting that each domain may have internal disulfide bonds and that punctin consists of a series of independently folded and disulfide-bonded domains.
Specifications
| ADAMTSL1 | |
| Polyclonal | |
| Unconjugated | |
| ADAMTSL1 | |
| 5930437A14Rik; 6720426B09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 1; ADAM-TS related protein 1; Adamtsl1; ADAMTSL-1; ADAMTS-like 1; ADAMTS-like protein 1; ADAMTSR1; C9orf94; DKFZp686L03130; FLJ35283; FLJ41032; FLJ46891; metalloendopeptidases; MGC118803; MGC118805; MGC40193; PUNCTIN; Punctin-1; RP11-220B22.2; UNQ528/PRO1071 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92949 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8N6G6 | |
| ADAMTSL1 | |
| Recombinant protein encompassing a sequence within the center region of human ADAMTSL1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction