Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTSL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562847 Supplier Invitrogen™ Supplier No. PA562847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VMSAYAMCVR YDGVEVDDSY CDALTRPEPV HEFCAGRECQ PRWETSSWSE CS Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 100%.

ADAMTSL2, or A Disintegrin And Metalloproteinase with Thrombospondin Motifs Like 2, is a gene that codes for a secreted glycoprotein involved in the regulation of extracellular matrix (ECM) and associated proteases. The gene, initially identified by the Kazusa DNA Research Institute, plays a crucial role in various biological processes including ECM remodeling. Mutations or alterations in ADAMTSL2 are implicated in multiple diseases, including genetic disorders like geleophysic dysplasia and cancer. Its expression levels and genetic variants have been correlated with cancer progression, immune cell infiltration in tumors, and other pathological conditions like non-alcoholic steatohepatitis (NASH). ADAMTSL2 is a potential biomarker for disease prognosis and therapeutic targeting due to its significant role in ECM dynamics and disease pathology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTSL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAMTSL2
Gene Accession No. Q86TH1
Gene Alias a disintegrin and metalloproteinase; A930008K15Rik; ADAM; ADAMs; ADAMTS like 2; Adamtsl2; ADAMTSL-2; ADAMTS-like 2; ADAMTS-like protein 2; GPHYSD1; Kiaa0605; metalloendopeptidases; RGD1305459; Tcp1; TCP-1; TSP1-repeat-containing protein 1; TSP1-repeats containing protein; TSP1-repeats-containing protein 1
Gene Symbols ADAMTSL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAMTSL2. Recombinant protein control fragment (Product #RP-104491).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9719
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.