Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTSL4 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305785MFV500
Description
ADAMTSL4 Polyclonal antibody specifically detects ADAMTSL4 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
ADAMTSL4 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
ADAMTSL-4, ADAMTS-like 4, PP1396, thrombospondin repeat containing 1, TSRC1 | |
Produced in rabbits immunized with E. coli-derived Human ADAMTSL4 fragment. | |
0.1 mL | |
Apoptosis, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
54507 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction