Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTSL4 Monoclonal Antibody (2H7)
GREENER_CHOICE

Catalog No. PIMA524477
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524477 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524477 Supplier Invitrogen™ Supplier No. MA524477
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MPAPPHPRTP LGSPAAYWKR VGHSACSASC GKGVWRPIFL CISRESGEEL DERSCAAGAR PPASPEPCHG TPCPPYWEAG EWTSCSRSCG PGTQHRQLQC RQEFGGGGSS VPPERCGHLP RPNITQSCQL RLCGHWEVGS PWSQCSVRCG RGQRSRQVRC VGNNGDEVSE QECASGPPQP PSREACDMGP CTTAWFHSDW SSKCSAECGT GIQRRSVVCL GSGAALGPGQ GEAGAGTGQS CPTGSRPPDM RACSLGPCER TWRWYTGPWG ECSSECGSGT QRRDIICVSK LGTEFNVTSP SNCSHLPRPP ALQPCQGQAC QDRWFSTPWS PCSRSCQGGT QTREVQCLST NQTLSTRCPP QLRPSRKRPC NSQPCSQRPD DQCKDSSPHC PLVVQARLCV YPYYTATCCR SCAHVLERSP QDPS.

This gene is a member of ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs)-like gene family and encodes a protein with protein with seven thrombospondin type 1 repeats. The thrombospondin type 1 repeat domain is found in many proteins with diverse biological functions including cellular adhesion, angiogenesis, and patterning of the developing nervous system. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTSL4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2H7
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ADAMTSL4
Gene Accession No. Q6UY14
Gene Alias a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 4; adamtsl4; ADAMTSL-4; adamtsl4 {ECO:0000312; ADAMTS-like 4; ADAMTS-like protein 4; ECTOL2; EMBL:AAH99119.1}; metalloendopeptidases; PP1396; thrombospondin repeat containing 1; thrombospondin repeat protein 1; thrombospondin repeat-containing protein 1; Tsrc1; UNQ2803/PRO34012
Gene Symbols ADAMTSL4
Host Species Mouse
Immunogen ADAMTSL4 (AAH27478.2, 1 a.a. ~ 424 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54507
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.