Learn More
ADAMTSL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 153041AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0137
Only null left
Add to Cart
Description
Description
This gene is a member of ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs)-like gene family and encodes a protein with protein with seven thrombospondin type 1 repeats. The thrombospondin type 1 repeat domain is found in many proteins with diverse biological functions including cellular adhesion, angiogenesis, and patterning of the developing nervous system. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
ADAMTSL4
Polyclonal
Unconjugated
ADAMTSL4
ADAMTS like 4, ADAMTS like protein 4, ADAMTSL 4, ADAMTSL4, TSRC1, UNQ2803/PRO34012
Rabbit
Antigen Affinity Chromatography
RUO
229595, 310670, 54507
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4FZU4, Q6UY14, Q80T21
Adamtsl4
ADAMTSL4 Fusion Protein Ag7781
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.