Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTSL4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986867
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6867 Supplier Bioss Supplier No. BS5864R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs)-like gene family and encodes a protein with protein with seven thrombospondin type 1 repeats. The thrombospondin type 1 repeat domain is found in many proteins with diverse biological functions including cellular adhesion, angiogenesis, and patterning of the developing nervous system. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Specifications

Antigen ADAMTSL4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADAMTSL4
Gene Accession No. Q6UY14
Gene Alias a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 4; Adamtsl4; ADAMTSL-4; adamtsl4 {ECO:0000312; ADAMTS-like 4; ADAMTS-like protein 4; ECTOL2; EMBL:AAH99119.1}; metalloendopeptidases; PP1396; thrombospondin repeat containing 1; thrombospondin repeat protein 1; Thrombospondin repeat-containing protein 1; TSRC1; UNQ2803/PRO34012
Gene Symbols ADAMTSL4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ADAMTSL4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54507
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.