Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTSL5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP19343825UL
Description
ADAMTSL5 Polyclonal specifically detects ADAMTSL5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ADAMTSL5 | |
Polyclonal | |
Western Blot 1:100-1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
Q6ZMM2 | |
ADAMTSL5 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AHRLLHYCGSDFVFQARVLGHHHQAQETRYEVRIQLVYKNRSPLRAREYVWAPGHCPCPMLAPHRDYLMAVQRLVSPDGTQDQ | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ADAMTSL-5, ADAMTS-like 5, ADAMTS-like protein 5, thrombospondin type-1 domain-containing protein 6, thrombospondin, type I, domain containing 6, THSD6 | |
Rabbit | |
Affinity Purified | |
RUO | |
339366 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction