Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTSL5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560604
Description
Immunogen sequence: AHRLLHYCGS DFVFQARVLG HHHQAQETRY EVRIQLVYKN RSPLRAREYV WAPGHCPCPM LAPHRDYLMA VQRLVSPDGT QDQ Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 26%.
ADAMTSL5 belongs to a large superfamily containing 20 ADAMTS proteases and at least eight ADAMTS-like proteins. ADAMTS proteases are secreted enzymes with a conserved organization that includes a metalloprotease domain and an ancillary domain containing one or more thrombospondin type 1 repeats (TSR). The ADAMTS-like subfamily comprises proteins homologous to the ADAMTS ancillary domains but lacking the protease domain and hence lacking catalytic activity. The exact role of ADAMTSL5 is currently unknown, but other ADAMTS-like proteins appear to have regulatory roles in the extracellular matrix.
Specifications
ADAMTSL5 | |
Polyclonal | |
Unconjugated | |
ADAMTSL5 | |
2010109H09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 5; ADAMTSL5; ADAMTSL-5; ADAMTS-like 5; ADAMTS-like protein 5; metalloendopeptidases; Thrombospondin type-1 domain-containing protein 6; thrombospondin, type I domain containing 6; thrombospondin, type I, domain containing 6; THSD6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
339366 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZMM2 | |
ADAMTSL5 | |
Recombinant protein corresponding to Human ADAMTSL5. Recombinant protein control fragment (Product #RP-99196). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction