Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTSL5 Polyclonal Antibody

Chicken Polyclonal Antibody

Supplier:  Invitrogen™ PA520934

Catalog No. PIPA520934


Only null left
Add to Cart

Description

Description

A suggested positive control is human skeletal muscle tissue lysate. PA5-20934 can be used with blocking peptide PEP-1048.

ADAMTSL5 belongs to a large superfamily containing 20 ADAMTS proteases and at least eight ADAMTS-like proteins. ADAMTS proteases are secreted enzymes with a conserved organization that includes a metalloprotease domain and an ancillary domain containing one or more thrombospondin type 1 repeats (TSR). The ADAMTS-like subfamily comprises proteins homologous to the ADAMTS ancillary domains but lacking the protease domain and hence lacking catalytic activity. The exact role of ADAMTSL5 is currently unknown, but other ADAMTS-like proteins appear to have regulatory roles in the extracellular matrix.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ADAMTSL5
Polyclonal
Unconjugated
ADAMTSL5
2010109H09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 5; ADAMTSL5; ADAMTSL-5; ADAMTS-like 5; ADAMTS-like protein 5; metalloendopeptidases; Thrombospondin type-1 domain-containing protein 6; thrombospondin, type I domain containing 6; thrombospondin, type I, domain containing 6; THSD6
Chicken
Antigen affinity chromatography
RUO
314626, 339366, 66548
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q6ZMM2
ADAMTSL5
A 17 amino acid peptide near the carboxy terminus of human ADAMTSL5.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.