Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529331 Supplier Invitrogen™ Supplier No. PA529331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Jurkat, Raji. Predicted reactivity: Mouse (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The adhesion and degranulation adaptor protein (ADAP) was initially identified as a molecular adapter that couples T cell receptor (TCR) stimulation to the avidity of integrins governing T cell adhesion. TCR stimulation promotes the formation of a multi-protein complex containing CARMA1, MALT1, and BCL-10, which through the association of ADAP, ultimately activates the NF-kappa-B family of transcription factors. More recent experiments have shown that ADAP controls optimal T cell proliferation, cytokine production, and expression of the Bcl-2 family member Bcl-x(L), suggesting that ADAP regulates T cell activation by promoting antigen-dependent T cell-antigen presenting cell (APC) activation. At least three isoforms of ADAP are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAP
Applications Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene FYB1
Gene Accession No. O15117
Gene Alias ADAP; adhesion and degranulation promoting adaptor protein; adhesion and degranulation-promoting adaptor protein; B630013F22Rik; Fyb; Fyb1; FYB-120/130; FYN binding protein; FYN binding protein (FYB-120/130); FYN binding protein FYB-130; FYN-binding protein; FYN-binding protein 1; FYN-T-binding protein; p120/130; p120/p130; PRO0823; RGD1563421; Slap130; SLAP-130; SLP-76-associated phosphoprotein
Gene Symbols FYB1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FYB. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2533
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.