Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAP Recombinant Rabbit Monoclonal Antibody (JE44-80)
GREENER_CHOICE

Catalog No. PIMA534840
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534840 Supplier Invitrogen™ Supplier No. MA534840
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: U937, human tonsil tissue, human appendix tissue, human spleen tissue.

The adhesion and degranulation adaptor protein (ADAP) was initially identified as a molecular adapter that couples T cell receptor (TCR) stimulation to the avidity of integrins governing T cell adhesion. TCR stimulation promotes the formation of a multi-protein complex containing CARMA1, MALT1, and BCL-10, which through the association of ADAP, ultimately activates the NF-kappa-B family of transcription factors. More recent experiments have shown that ADAP controls optimal T cell proliferation, cytokine production, and expression of the Bcl-2 family member Bcl-x(L), suggesting that ADAP regulates T cell activation by promoting antigen-dependent T cell-antigen presenting cell (APC) activation. At least three isoforms of ADAP are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAP
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE44-80
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene FYB1
Gene Accession No. O15117
Gene Alias ADAP; adhesion and degranulation promoting adaptor protein; adhesion and degranulation-promoting adaptor protein; B630013F22Rik; Fyb; Fyb1; FYB-120/130; FYN binding protein; FYN binding protein (FYB-120/130); FYN binding protein FYB-130; FYN-binding protein; FYN-binding protein 1; FYN-T-binding protein; p120/130; p120/p130; PRO0823; RGD1563421; Slap130; SLAP-130; SLP-76-associated phosphoprotein
Gene Symbols FYB1
Host Species Rabbit
Immunogen Synthetic peptide within Human FYB aa 701-750.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2533
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.