Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 139111AP150UL
Description
Centaurin alpha-1 (ADAP1, ArfGAP With Dual PH Domains 1) is a Protein Coding gene. Among its related pathways are B Cell Receptor Signaling Pathway (sino) and Arf6 signaling events. Gene Ontology (GO) annotations related to this gene include GTPase activator activity and inositol 1,3,4,5 tetrakisphosphate binding. An important paralog of this gene is ADAP2.Specifications
| ADAP1 | |
| Polyclonal | |
| Unconjugated | |
| ADAP1 | |
| ADAP1, ArfGAP with dual PH domains 1, CENTA1, Centaurin alpha 1, Cnt a1, GCS1L, p42IP4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11033, 171097, 231821 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75689 | |
| Adap1 | |
| ADAP1 Fusion Protein Ag4898 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction