Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Adaptaquin
GREENER_CHOICESDP

Catalog No. 595310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-531-0 10 mg
59-535-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-531-0 Supplier Tocris Bioscience™ Supplier No. 5953/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

HIF-prolyl hydroxylase-2 (PHD2) inhibitor; antioxidant

Adaptaquin is a HIF-prolyl hydroxylase-2 (PHD2) inhibitor. Blocks glutamate induced ROS production in HT-22 cells, independent of MnSOD. Exhibits neuroprotective effects and enhances functional recovery in rodent intracerebral hemorrhage models, via inhibition of ATF4 dependent genes. Brain penetrant. Also antioxidant.

Specifications

Chemical Name or Material 7-[(4-Chlorophenyl)[(3-hydroxy-2-pyridinyl)amino]methyl]-8-quinolinol
CAS 385786-48-1
Quantity 10 mg
Target Hydrolase Inhibitors
Molecular Formula C21H16ClN3O2
Formula Weight 377.82
Purity >97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.