Description
- For Rotor F-35-6-30
Specifications
Specifications
| Type | Adapter |
| Tube Type | Round Bottom |
| Max. Tube Length | 65 to 89 mm |
| For Use With (Equipment) | for Rotor F-35-6-30, small rotor bore |
| No. of Places | 12 |