Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Catalog No. p-7078636
Click to view available options
For Use With (Equipment):
for Rotor F-35-6-30, large rotor bore
for Rotor F-35-6-30, small rotor bore
Max. Tube Length:
65 to 89 mm
90 to 110 mm
Designed for use with Eppendorf Rotors and Centrifuges
  • For Rotor F-35-6-30
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Max. Tube Length 65 to 89 mm
For Use With (Equipment) for Rotor F-35-6-30, large rotor bore
No. of Places 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.