Learn More
Heidolph™ SQ13 Screwthread Adapters
Shop All Heidolph North America Products
Click to view available options
Type:
B19
B29
Description
- Adapter to SQ13 Screwthread
- Available in two different types
Specifications
Specifications
| For Use With (Equipment) | SQ13 Screwthread |
| For Use With (Application) | Allows for Connection of Gas or Vacuum |
| Quantity | 5/Pk. |
| Type | B19 |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov