Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Adapter for Conical Tubes

Catalog No. 05414325 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Rotor A-4-81 and Rotor FA-45-6-30
Rotor FA-45-6-30
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-414-325 Rotor FA-45-6-30
05-414-326 Rotor A-4-81 and Rotor FA-45-6-30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 05-414-325 Supplier Eppendorf™ Supplier No. 5820717009
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed for use with a variety of products to expand liquid handling capabilities in the lab

  • Available in packs of two or seven
  • For use with Eppendorf Rotors
TRUSTED_SUSTAINABILITY

Specifications

Type Adapter
Tube Type Conical
Capacity 15 mL
For Use With (Equipment) Rotor FA-45-6-30
Color Orange
No. per Pack Set of 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.