Description
- Suitable for S-4xUniversal rotor buckets
- Accommodates selected array of tubes, bottles, or plates
Specifications
Specifications
| Tube Type | Conical |
| Capacity | 9 x 50 mL Conical Tubes, 1 Microplate |
| For Use With (Equipment) | S-4xUniversal Rotor |
| No. of Places | 9/1 |
| No. per Pack | 2 |