Description
- Secures vessel in precise position
- Compatible with Q3 Sample Changer
- Conductive materials to reduce electrostatic charge build-up
- Highly accurate and safe filling
Specifications
Specifications
| For Use With (Equipment) | Q3 Sample Changer (30418662) |
| Size (Metric) | 13.5 x 20 mm |
| Type | Adapter Vial |