Learn More
Cytiva Adaptor Kit
Catalog No. 45002433 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Electrophoresis Systems
Catalog No. For Use With (Equipment)
45-002-433 Electrophoresis Systems
1 options
Catalog No. 45-002-433 Supplier Cytiva Supplier No. 80610527
Connects 4 mm output jack to 4 mm high voltage leads
TRUSTED_SUSTAINABILITY
Specifications
Description 4 mm Male/ 4 mm Female
For Use With (Application) Connecting 4 mm Output Jack to 4 mm High Voltage Leads
For Use With (Equipment) Electrophoresis Systems
Type Fixed Sleeve
Quantity 1 Kit