Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAR Mouse anti-Human, Clone:4E2E4, Abnova™

Catalog No. 89334358 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-358 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-358 Supplier Abnova Corporation Supplier No. MAB17910
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human ADAR.

This gene encodes the enzyme responsible for RNA editing by site-specific deamination of adenosines. This enzyme destabilizes double stranded RNA through conversion of adenosine to inosine. Mutations in this gene have been associated with dyschromatosis symmetrica hereditaria. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Specifications

Antigen ADAR
Applications ELISA, Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4E2E4
Conjugate Unconjugated
Description adenosine deaminase, RNA-specific
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunofluorescence (1:200-1:1000)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene ADAR
Gene Alias ADAR1, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88dsRBP, p136
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1085-1223 of human ADAR.
Quantity 100 μg
Gene ID (Entrez) 103
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.