Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAR1 Rat anti-Mouse, Human, Clone: RD4B11, MilliporeSigma™

Catalog No. MABE179025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1790-25 25 μg
MABE1790-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE1790-25 Supplier MilliporeSigma Supplier No. MABE179025UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-ADAR1, clone RD4B11 is a highly specific rat monoclonal antibody that targets ADAR1 and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAR1
Applications Western Blot
Classification Monoclonal
Clone RD4B11
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Adar
Host Species Rat
Immunogen His-tagged recombinant fragment corresponding to 401 amino acids from the internal region of murine Double-stranded RNA-specific adenosine deaminase (ADAR1). Immunogen sequnece is conserved isoforms 1 and 2.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_062629.3
Test Specificity Clone RD4B11 is a rat monoclonal antibody that detects Double-stranded RNA-specific adenosine deaminase in human and murine cells.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.