Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAR2 (Adenosine Deaminase Acting on RNA 2) Sheep anti-Rat, Polyclonal, Exalpha
SDP

Catalog No. 502101856
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1856 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1856 Supplier Vector Laboratories Supplier No. U115P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ADAR2 (Adenosine Deaminase Acting on RNA 2) Polyclonal antibody specifically detects ADAR2 (Adenosine Deaminase Acting on RNA 2) in Rat samples. It is validated for Western Blot

Specifications

Antigen ADAR2 (Adenosine Deaminase Acting on RNA 2)
Applications Western Blot
Classification Polyclonal
Concentration 0.5-2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide; pH
Gene ADARB1
Gene Accession No. P51400
Gene Alias 1700057H01Rik; ADAR2; Adarb1; adenosine deaminase, RNA specific B1; adenosine deaminase, RNA-specific, B1; adenosine deaminase, RNA-specific, B1 (homolog of rat RED1); adenosine deaminase, RNA-specific, B1 (RED1 homolog rat); AW124433; AW558573; BB220382; D10Bwg0447e; Double-stranded RNA-specific editase 1; DRABA2; DRADA2; dsRNA adenosine deaminase; dsRNA adenosine deaminase DRADA2; RED1; RED1 homolog; RNA editase; RNA editing deaminase 1; RNA editing deaminase of glutamate receptors; RNA-editing deaminase 1; RNA-editing enzyme 1
Host Species Sheep
Immunogen GST fusion protein containing aa 6-66 of the rat RED1 coding sequence
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25367
Target Species Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.