Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADARB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569102 Supplier Invitrogen™ Supplier No. PA569102
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 85%.

ADARB2 gene ontology annotations related to this gene include adenosine to inosine editing; RNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADARB2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADARB2
Gene Accession No. Q9NS39
Gene Alias Adar3; Adarb2; adenosine deaminase 3, RNA dependent; adenosine deaminase RNA specific B2 (inactive); adenosine deaminase, RNA specific B2 (inactive); adenosine deaminase, RNA-specific, B2; adenosine deaminase, RNA-specific, B2 (inactive); adenosine deaminase, RNA-specific, B2 (non-functional); adenosine deaminase, RNA-specific, B2 (RED1 homolog rat); adenosine deaminase, RNA-specific, B2 (RED2 homolog rat); double-stranded RNA-specific editase B2; dsRNA adenosine deaminase B2; homolog of rat BLUE; Red2; RED2 homolog; RNA dependent adenosine deaminase 3; RNA-dependent adenosine deaminase 3; RNA-editing deaminase 2; RNA-editing enzyme 2; si:dkey-255g15.1
Gene Symbols ADARB2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ADARB2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.