Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541836
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541836 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541836 Supplier Invitrogen™ Supplier No. PA541836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LFRSFQKLLS RIARDKWPHS LRVQKLDTYQ EYKEAASSYQ EAWSTLRKQV Sequence homology: Dog: 79%; Guinea Pig: 85%; Horse: 86%; Human: 100%; Mouse: 86%; Rat: 93%.

This gene is a member of the ADAR (adenosine deaminase acting on RNA) family. Using site-specific adenosine modification, proteins encoded by these genes participate in the pre-mRNA editing of nuclear transcripts. The protein encoded by this gene, tRNA-specific adenosine deaminase 1, is responsible for the deamination of adenosine 37 to inosine in eukaryotic tRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADAT1
Gene Accession No. Q9BUB4
Gene Alias Adat1; Adenosine deaminase; adenosine deaminase acting on tRNA; adenosine deaminase tRNA specific 1; adenosine deaminase, tRNA specific 1; adenosine deaminase, tRNA-specific 1; HADAT1; mADAT1; MMADAT1; tRNA-specific 1; tRNA-specific adenosine deaminase 1; tRNA-specific adenosine-37 deaminase; zgc:162299
Gene Symbols ADAT1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ADAT1 (aa 438-487).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23536
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.