Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583511
Description
Immunogen sequence: IAQLCYEHYG IRLPKKGKPE PNHEWTLLAA VVKIQSPADK ACDTPDKPVQ VTKEVVSMGT GTKCIGQSKM RKNGDILNDS HAEVIARRSF QR.
This gene is a member of the ADAR (adenosine deaminase acting on RNA) family. Using site-specific adenosine modification, proteins encoded by these genes participate in the pre-mRNA editing of nuclear transcripts. The protein encoded by this gene, tRNA-specific adenosine deaminase 1, is responsible for the deamination of adenosine 37 to inosine in eukaryotic tRNA.
Specifications
ADAT1 | |
Polyclonal | |
Unconjugated | |
ADAT1 | |
Adat1; Adenosine deaminase; adenosine deaminase acting on tRNA; adenosine deaminase tRNA specific 1; adenosine deaminase, tRNA specific 1; adenosine deaminase, tRNA-specific 1; HADAT1; mADAT1; MMADAT1; tRNA-specific 1; tRNA-specific adenosine deaminase 1; tRNA-specific adenosine-37 deaminase; zgc:162299 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23536 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUB4 | |
ADAT1 | |
Recombinant protein corresponding to Human ADAT1. Recombinant protein control fragment (Product #RP-98538). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction