Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557287
Description
Immunogen sequence: PKPAASGACS VSAEETEKWM EEAMHMAKEA LENTEVPVGC LMVYNNEVVG KGRNEVNQTK NATRHAEMVA IDQVLDWCRQ SGKSPSEVFE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 77%.
Probably participates in deamination of adenosine-34 to inosine in many tRNAs.
Specifications
ADAT2 | |
Polyclonal | |
Unconjugated | |
ADAT2 | |
4933426M09Rik; ADAT2; adenosine deaminase tRNA specific 2; adenosine deaminase, tRNA specific 2; adenosine deaminase, tRNA-specific 2; adenosine deaminase, tRNA-specific 2, TAD2 homolog; deadc1; deaminase domain containing 1; deaminase domain-containing protein 1; dJ20N2; dJ20N2.1; LOW QUALITY PROTEIN: tRNA-specific adenosine deaminase 2; si:ch211-157l15.2; TAD2; tRNA-specific adenosine deaminase 2; tRNA-specific adenosine deaminase 2 homolog; tRNA-specific adenosine-34 deaminase subunit ADAT2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
134637 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6V5 | |
ADAT2 | |
Recombinant protein corresponding to Human ADAT2. Recombinant protein control fragment (Product #RP-95034). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction