Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570759
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570759 Supplier Invitrogen™ Supplier No. PA570759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%; Zebrafish: 92%.

Probably participates in deamination of adenosine-34 to inosine in many tRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADAT2
Gene Accession No. Q7Z6V5
Gene Alias 4933426M09Rik; ADAT2; adenosine deaminase tRNA specific 2; adenosine deaminase, tRNA specific 2; adenosine deaminase, tRNA-specific 2; adenosine deaminase, tRNA-specific 2, TAD2 homolog; deadc1; deaminase domain containing 1; deaminase domain-containing protein 1; dJ20N2; dJ20N2.1; LOW QUALITY PROTEIN: tRNA-specific adenosine deaminase 2; si:ch211-157l15.2; TAD2; tRNA-specific adenosine deaminase 2; tRNA-specific adenosine deaminase 2 homolog; tRNA-specific adenosine-34 deaminase subunit ADAT2
Gene Symbols ADAT2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human ADAT2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134637
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.