Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114193 Supplier Invitrogen™ Supplier No. PA5114193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPFPACSFAP AAAPQAVRAG AVRKLDADED GLPYLCTGYD LYVTREPCAM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.

This gene encodes a subunit of a tRNA-specific adenosine deaminase. This heterodimeric enzyme converts adenosine to inosine in the tRNA anticodon. A mutation in this gene causes a syndrome characterized by intellectual disability and strabismus. This gene shares its 5' exon with the overlapping gene, secretory carrier membrane protein 4. ADAT3 (Adenosine Deaminase TRNA Specific 3) is a Protein Coding gene. Diseases associated with ADAT3 include Neurodevelopmental Disorder With Brain Abnormalities, Poor Growth, And Dysmorphic Facies and Cone-Rod Dystrophy 10. Among its related pathways are tRNA processing and Processing of Capped Intron-Containing Pre-mRNA. Gene Ontology (GO) annotations related to this gene include hydrolase activity. An important paralog of this gene is ADAT2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAT3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADAT3
Gene Accession No. Q96EY9
Gene Alias ADAT3; adenosine deaminase, tRNA specific 3; adenosine deaminase, tRNA-specific 3; adenosine deaminase, tRNA-specific 3, TAD3 homolog; FWP005; MRT36; MST121; MSTP121; Probable inactive tRNA-specific adenosine deaminase-like protein 3; S863-5; TAD3; tRNA-specific adenosine deaminase 3 homolog; tRNA-specific adenosine deaminase-like protein 3; tRNA-specific adenosine-34 deaminase subunit ADAT3
Gene Symbols ADAT3
Host Species Rabbit
Immunogen Immunogen sequence: RPFPACSFAP AAAPQAVRAG AVRKLDADED GLPYLCTGYD LYVTREPCAM (aa 244-293).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113179
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.