Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADCK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528685
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528685 Supplier Invitrogen™ Supplier No. PA528685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (87%), Rat (85%), Rhesus Monkey (98%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ADCK1 is a protein coding gene. The function of this protein is not yet clear. It is not known if it has protein kinase activity and what type of substrate it would phosphorylate (Ser, Thr or Tyr).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADCK1
Applications Western Blot
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ADCK1
Gene Accession No. Q86TW2
Gene Alias 2610005A10Rik; aarF domain containing kinase 1; AarF domain-containing protein kinase 1; Adck1; Nrbf1; uncharacterized aarF domain-containing protein kinase 1
Gene Symbols ADCK1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ADCK1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57143
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.