Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADCK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101195 Supplier Invitrogen™ Supplier No. PA5101195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ADCK1.

ADCK1 is a protein coding gene. The function of this protein is not yet clear. It is not known if it has protein kinase activity and what type of substrate it would phosphorylate (Ser, Thr or Tyr).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADCK1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADCK1
Gene Accession No. Q86TW2, Q9D0L4
Gene Alias 2610005A10Rik; aarF domain containing kinase 1; AarF domain-containing protein kinase 1; Adck1; Nrbf1; uncharacterized aarF domain-containing protein kinase 1
Gene Symbols ADCK1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADCK1(Accession Q86TW2), corresponding to amino acid residues V267-H317.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57143, 72113
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.