Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADCK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101193
Description
Antibody detects endogenous levels of total ADCK2.
The function of this protein is not yet clear. It is not known if it has protein kinase activity and what type of substrate it would phosphorylate (Ser, Thr or Tyr).
Specifications
ADCK2 | |
Polyclonal | |
Unconjugated | |
ADCK2 | |
AA408112; AARF; aarF domain containing kinase 2; Adck2; BB006057; MGC20727; putative ubiquinone biosynthesis protein AarF; uncharacterized aarF domain-containing protein kinase 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
312258, 90956 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7Z695 | |
ADCK2 | |
A synthesized peptide derived from human ADCK2(Accession Q7Z695), corresponding to amino acid residues S236-S286. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction