Learn More
Invitrogen™ ADCK2 Polyclonal Antibody

Description
ADCK2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | ADCK2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ADCK2 |
| Gene Accession No. | Q6NSR3 |
| Gene Alias | AA408112; AARF; aarF domain containing kinase 2; Adck2; BB006057; MGC20727; putative ubiquinone biosynthesis protein AarF; uncharacterized aarF domain-containing protein kinase 2 |
| Gene Symbols | ADCK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.