Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADCK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106505
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106505 Supplier Invitrogen™ Supplier No. PA5106505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ADCK5.

ADCK5 is member of the AarF domain containing kinase family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADCK5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADCK5
Gene Accession No. Q3MIX3, Q80V03
Gene Alias A230108P17; aarF domain containing kinase 5; Adck5; Uncharacterized aarF domain-containing protein kinase 5
Gene Symbols ADCK5
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADCK5(Accession Q3MIX3), corresponding to amino acid residues P45-L95.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203054, 268822
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.