Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCK5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988199
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8199 Supplier Bioss Supplier No. BS8012R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADCK5 is member of the AarF domain containing kinase family.

Specifications

Antigen ADCK5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADCK5
Gene Accession No. Q3MIX3
Gene Alias A230108P17; aarF domain containing kinase 5; ADCK5; Uncharacterized aarF domain-containing protein kinase 5
Gene Symbols ADCK5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ADCK5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203054
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.