Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985578
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5578 Supplier Bioss Supplier No. BS3681R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADCY1 belongs to the adenylyl cyclase class-4/guanylyl cyclase family. It is a calmodulin-sensitive adenylyl cyclase. ADCY1 may be involved in regulatory processes in the central nervous system. It may play a role in memory acquisition and learning.

Specifications

Antigen ADCY1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADCY1
Gene Accession No. Q08828
Gene Alias 3′,5′-cyclic AMP synthetase; AC1; ADCY1; adenyl cyclase; adenylate cyclase 1; adenylate cyclase 1 (brain); Adenylate cyclase type 1; Adenylate cyclase type I; Adenylyl cyclase 1; ATP pyrophosphate-lyase 1; barrelless; brl; Ca(2+)/calmodulin-activated adenylyl cyclase; D11Bwg1392e; DFNB44; I-AC; mKIAA4070
Gene Symbols ADCY1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Adenylyl cyclase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.